Merck
  • Pathological vitreous causes cell line-derived (but not donor-derived) retinal pigment epithelial cells to display proliferative vitreoretinopathy-like features in culture.
Pathological vitreous causes cell line-derived (but not donor-derived) retinal pigment epithelial cells to display proliferative vitreoretinopathy-like features in culture.
Clinical & experimental ophthalmology (2014-03-13)
Maryada Sharma, Anil Tiwari, Shweta Sharma, Reema Bansal, Vishali Gupta, Amod Gupta, Manni Luthra-Guptasarma
ZUSAMMENFASSUNG
It is well understood that epithelial mesenchymal transformation occurs when retinal pigment epithelial cells, sourced from either a cell line or cadaver eye, are cultured in the presence of cadaver-derived vitreous. We sought to study the changes in retinal pigment epithelial cells when cell line-derived retinal pigment epithelial cells are cultured in the presence of pathological vitreous. Prospective study. 42 patients with rhegmatogenous retinal detachments. D407 retinal pigment epithelial cells were cultured in the presence of cadaver-derived vitreous or vitreous/subretinal fluid derived from patients undergoing retinal reattachment surgeries. Besides the changes in phenotypic characteristics, the viability, proliferation, migration, mesenchymal marker expression and changes in the extracellular matrix components were also evaluated. Fibrotic phenotype in cell culture. Our study clearly demonstrates that cell line-derived retinal pigment epithelial cells (unlike donor-derived retinal pigment epithelial cells) cultured in the presence of patient-derived vitreous/subretinal fluid, exhibit characteristic features of proliferative vitreoretinopathy. We propose that it is the synergistic effect of the combined use of (i) pathological vitreous, rather than cadaver-derived vitreous (since rhegmatogenous retinal detachment-derived pathological vitreous and subretinal fluid contain exaggerated amounts of growth factors, which could predispose to proliferative vitreoretinopathy development) and (ii) cells from an immortal cell culture (cell line), rather than from primary cell cultures (since cells subjected to continuous serial passaging acquire some mesenchymal characteristics), which together result in not only a unique phenotype, but also prime these cells towards display of features associated with proliferative vitreoretinopathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Monoklonaler Anti-Kollagen-Antikörper, Typ I in Maus hergestellte Antikörper, clone COL-1, ascites fluid
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Anti-Kollagen-Typ-I-Antikörper, Klon 5D8-G9, clone 5D8-G9, Chemicon®, from mouse