Merck
  • Click synthesis of glucose-functionalized hydrophilic magnetic mesoporous nanoparticles for highly selective enrichment of glycopeptides and glycans.
Click synthesis of glucose-functionalized hydrophilic magnetic mesoporous nanoparticles for highly selective enrichment of glycopeptides and glycans.
Journal of chromatography. A (2014-07-22)
Jiangnan Zheng, Yun Xiao, Ling Wang, Zian Lin, Huanghao Yang, Lan Zhang, Guonan Chen
ZUSAMMENFASSUNG
Selective enrichment of glycopeptides from complex biological samples is essential for MS-based glycoproteomics, but challenges still remain. In this work, glucose-functionalized magnetic mesoporous nanoparticles (MMNs), which hold the attractive features of well-defined core/shell structure, high specific surface area (324m(2)/g), narrow pore size distribution (2.2nm) and high magnetic responsivity (69.1emug(-1)), were synthesized via click chemistry and applied to enrich glycopeptides and glycans. Taking advantages of the size-exclusive effect of mesopore against proteins and the hydrophilic interaction between glycans and glucose, the hydrophilic MMNs possessed high selectivity for glycopeptides at the digested mixtures of horseradish peroxidase (HRP), myoglobin and β-casein at molar ratio of 1:1:10, large enrichment capacity (as high as 250mg/g), high sensitivity (50fmol), excellent speed (5min for enrichment) and high recovery of glycopeptides (as high as 94.6%). Additionally, the MMNs exhibited excellent performance in enrichment of N-linked glycans from the digested human serum that are made up of peptides, large proteins and other compounds. These outstanding features will give the hydrophilic MMNs high benefit for MS analysis of low-abundance glycopeptides/glycans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylenglycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
(+)-Natrium-L-ascorbat, crystalline, ≥98%
USP
Ethylenglycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylenglycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-Natrium-L-ascorbat, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
Myoglobin aus Pferdeherz, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
β-Kasein aus Kuhmilch, BioUltra, ≥98% (PAGE)
Supelco
Natriumzitrat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylenglycol, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)