Merck
The expression of p21 is upregulated by forkhead box A1/2 in p53-null H1299 cells.
FEBS letters (2014-10-05)
Joo-Hee An, Sang-Min Jang, Jung-Woong Kim, Chul-Hong Kim, Peter I Song, Kyung-Hee Choi
ZUSAMMENFASSUNG
The expression of the cell cycle inhibitor p21 is increased in response to various stimuli and stress signals through p53-dependent and independent pathways. We demonstrate in this study that forkhead box A1/2 (FOXA1/2) is a crucial transcription factor in the activation of p21 transcription via direct binding to the p21 promoter in p53-null H1299 lung carcinoma cells. In addition, histone deacetylase inhibitor trichostatin A (TSA)-mediated upregulation of p21 expression was repressed by knockdown of FOXA1/2 in H1299 cells. Consequently, these results suggest that FOXA1/2 is required for p53-independent p21 expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)