Merck
  • Synthesis of a red fluorescent dye-conjugated Ag@SiO2 nanocomposite for cell immunofluorescence.
Synthesis of a red fluorescent dye-conjugated Ag@SiO2 nanocomposite for cell immunofluorescence.
Applied spectroscopy (2015-01-15)
Meicong Dong, Yu Tian, Dimitri Pappas
ZUSAMMENFASSUNG
In this work we describe a one-step approach for incorporating a red fluorophore (2SBPO) into core-shell nanoparticles for metal-enhanced fluorescence immunolabels. The 2SBPO-MEF nanoparticles are particularly attractive as cell labels because their ∼ 670 nm emission has minimal overlap with cell autofluorescence and from overlap with many conventional probes. 2SBPO was incorporated through physical entrapment during the Stöber process. Antibody-based cell labels were then synthesized using covalent linkage. The nanoparticle fluorescence was 7.5-fold higher than control nanoparticles lacking a metal core. We demonstrated labeling of CD4 + HuT 78 T lymphocytes using anti-CD4-conjugated nanoparticle labels. Cells labeled with anti-CD4 nanoparticles showed a 35-fold fluorescence signal compared to anti-CD4 coreless controls. This simple synthesis protocol can be applied to a variety of hydrophilic fluorophore types and has broad potential in bioanalytical and biosensing applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ammoniumhydroxid -Lösung, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Natriumzitrat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexancarbonsäure, 98%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O