Merck
  • Generation of insulin-producing β-like cells from human iPS cells in a defined and completely xeno-free culture system.
Generation of insulin-producing β-like cells from human iPS cells in a defined and completely xeno-free culture system.
Journal of molecular cell biology (2014-06-28)
Hussain Md Shahjalal, Nobuaki Shiraki, Daisuke Sakano, Kazuhide Kikawa, Soichiro Ogaki, Hideo Baba, Kazuhiko Kume, Shoen Kume
ZUSAMMENFASSUNG
Human induced pluripotent stem (hiPS) cells are considered a potential source for the generation of insulin-producing pancreatic β-cells because of their differentiation capacity. In this study, we have developed a five-step xeno-free culture system to efficiently differentiate hiPS cells into insulin-producing cells in vitro. We found that a high NOGGIN concentration is crucial for specifically inducing the differentiation first into pancreatic and duodenal homeobox-1 (PDX1)-positive pancreatic progenitors and then into neurogenin 3 (NGN3)-expressing pancreatic endocrine progenitors, while suppressing the differentiation into hepatic or intestinal cells. We also found that a combination of 3-isobutyl-1-methylxanthine (IBMX), exendin-4, and nicotinamide was important for the differentiation into insulin single-positive cells that expressed various pancreatic β-cell markers. Most notably, the differentiated cells contained endogenous C-peptide pools that were released in response to various insulin secretagogues and high levels of glucose. Therefore, our results demonstrate the feasibility of generating hiPS-derived pancreatic β-cells under xeno-free conditions and highlight their potential to treat patients with type 1 diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
SAFC
L-Glutamin
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Carbamoylcholinchlorid, ≥98% (titration), crystalline
Sigma-Aldrich
TGF-β RI Kinase-Inhibitor VI, SB431542, CAS-Nr. 301836-41-9, Calbiochem, TGF-β RI Kinase Inhibitor VI, SB431542, CAS 301836-41-9, is a cell-permeable inhibitor of SMAD2 phosphorylation. Inhibits the activity of ALK4 and ALK5 (IC50 = 140 nM and 94 nM, respectively).
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Niacinamid, meets USP testing specifications
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
Tolbutamid, analytical standard
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98.5% (HPLC)
Nicotinamid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Carbachol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Tolbutamid, European Pharmacopoeia (EP) Reference Standard