Merck
Bmi1 promotes erythroid development through regulating ribosome biogenesis.
Stem cells (Dayton, Ohio) (2014-11-12)
Rui Gao, Sisi Chen, Michihiro Kobayashi, Hao Yu, Yingchi Zhang, Yang Wan, Sara K Young, Anthony Soltis, Ming Yu, Sasidhar Vemula, Ernest Fraenkel, Alan Cantor, Yevgeniy Antipin, Yang Xu, Mervin C Yoder, Ronald C Wek, Steven R Ellis, Reuben Kapur, Xiaofan Zhu, Yan Liu
ZUSAMMENFASSUNG
While Polycomb group protein Bmi1 is important for stem cell maintenance, its role in lineage commitment is largely unknown. We have identified Bmi1 as a novel regulator of erythroid development. Bmi1 is highly expressed in mouse erythroid progenitor cells and its deficiency impairs erythroid differentiation. BMI1 is also important for human erythroid development. Furthermore, we discovered that loss of Bmi1 in erythroid progenitor cells results in decreased transcription of multiple ribosomal protein genes and impaired ribosome biogenesis. Bmi1 deficiency stabilizes p53 protein, leading to upregulation of p21 expression and subsequent G0/G1 cell cycle arrest. Genetic inhibition of p53 activity rescues the erythroid defects seen in the Bmi1 null mice, demonstrating that a p53-dependent mechanism underlies the pathophysiology of the anemia. Mechanistically, Bmi1 is associated with multiple ribosomal protein genes and may positively regulate their expression in erythroid progenitor cells. Thus, Bmi1 promotes erythroid development, at least in part through regulating ribosome biogenesis. Ribosomopathies are human disorders of ribosome dysfunction, including Diamond-Blackfan anemia (DBA) and 5q- syndrome, in which genetic abnormalities cause impaired ribosome biogenesis, resulting in specific clinical phenotypes. We observed that BMI1 expression in human hematopoietic stem and progenitor cells from patients with DBA is correlated with the expression of some ribosomal protein genes, suggesting that BMI1 deficiency may play a pathological role in DBA and other ribosomopathies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Chromatin-Immunpräzipitations-Kit EZ-Magna ChIP® A/G, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads. Control primers included.
Sigma-Aldrich
Anti-trimethyl-Histon H3-(Lys4-)Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Vinylencarbonat, contains ≤2% BHT as stabilizer, 97%