Merck
  • Nuclear expression of phosphorylated focal adhesion kinase is associated with poor prognosis in human colorectal cancer.
Nuclear expression of phosphorylated focal adhesion kinase is associated with poor prognosis in human colorectal cancer.
Anticancer research (2014-07-31)
Abdulkader Albasri, Wakkas Fadhil, John H Scholefield, Lindy G Durrant, Mohammad Ilyas
ZUSAMMENFASSUNG
To determine whether phosphorylated focal adhesion kinase (P-FAK) has prognostic value in colorectal cancer (CRC) and to test whether it has any association with Tensin 4 (TNS4) expression. P-FAK expression was assessed using immunohistochemistry in 462 CRC cases arrayed on a tissue microarray. P-FAK and TNS4 expression were assessed by immunohistochemistry in 40 cases of paired primary colorectal cancer and corresponding hepatic metastases. Nuclear P-FAK expression was observed in 44% of studied cases. Positive nuclear P-FAK expression was associated with shorter disease-specific survival in univariate (p=0.005) and multivariate analysis (p=0.016). P-FAK expression was greater in metastases than the primary tumours (p<0.001) and showed significant association with nuclear TNS4 (p<0.001) in metastases. P-FAK expression is an independent prognostic marker in CRC. The present data suggest that the FAK signalling pathway may interact with TNS4, a known oncogene in CRC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.