Merck
  • Genetic analysis of the human Insulin-like 3 gene: absence of mutations in a Greek paediatric cohort with testicular maldescent.
Genetic analysis of the human Insulin-like 3 gene: absence of mutations in a Greek paediatric cohort with testicular maldescent.
Andrologia (2015-03-03)
C Mamoulakis, I Georgiou, F Dimitriadis, P Tsounapi, I Giannakis, A Chatzikyriakidou, S Antypas, F Sofras, A Takenaka, N Sofikitis
ZUSAMMENFASSUNG
This study investigated the hypothesis that genetic alterations of the human insulin-like 3 (INSL3) gene are associated with testicular maldescent (TMD). Genomic DNA was extracted and amplified from peripheral blood samples of 170 unrelated children with all possible phenotypical expressions of TMD and 50 volunteers with normal external genitalia from the general paediatric population (controls). PCR-single strand conformation polymorphism analysis was used to screen INSL3 gene for genetic variants. For rapid screening of a detected nonsilent genetic alteration, restriction assay using endonuclease Eag I was further employed. Products were analysed on 2% agarose gel and restriction patterns were visualised by ethidium bromide. Differences in genotype and allelic distributions of nonsilent genetic alterations were evaluated between (i) patients-controls, (ii) familial-sporadic, (iii) bilateral-unilateral and (iv) intra-abdominal-inguinal cases of TMD. No mutations were detected. Three common INSL3 gene polymorphisms (27G>A, 126G>A, 178G>A) unrelated to any particular phenotype of TMD were detected both in patients and controls. These results indicate that INSL3 gene mutations are not a common cause of TMD in the human.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O