Merck
  • Effects of neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) treatment on ovarian development of the sapphire devil, Chrysiptera cyanea.
Effects of neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) treatment on ovarian development of the sapphire devil, Chrysiptera cyanea.
Fish physiology and biochemistry (2014-11-05)
Muhammad Badruzzaman, Satoshi Imamura, Yuki Takeuchi, Taro Ikegami, Akihiro Takemura
ZUSAMMENFASSUNG
In the neuroendocrine system controlling fish reproduction, dopamine (DA) acts as a gonadotropin inhibitory factor and plays a role in regulating gonadal development of certain species. The present study examined the effects of chemical destruction of dopaminergic neurons in the brain on DA production and ovarian development in the sapphire devil Chrysiptera cyanea, a reef-associated damselfish. The avidin-biotin-peroxidase complex method using an antibody against tyrosine hydroxylase (TH), a critical enzyme in the DA synthesis pathway, identified a population of dopaminergic neurons with somata in the anteroventral preoptic nucleus of the diencephalon and fibers terminating in the proximal pars distalis of the pituitary. Maintaining fish in seawater containing 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) at 0.02 and 0.2 µg/mL for 2 days resulted in decreases in DA, 3,4-dihydroxyphenylacetic acid (DOPAC; DA metabolite), and DA metabolic rate in the whole brain. The number of TH-positive neurons in the diencephalon decreased after 0.02 µg/mL MPTP treatment for 2 days. These results suggest that MPTP treatment destroys TH-positive neurons in the diencephalon, thereby decreasing the synthesis and release of DA from the brain. This treatment rescued ovarian development in fish with artificially retracted ovaries during the spawning season. The gonadosomatic index of MPTP-treated fish 5 and 7 days after treatment was significantly higher than that of control fish. Oocytes in the vitellogenic stages were observed in the ovaries of MPTP-treated fish, but not in control fish. These results suggest that DA in the brain drives ovarian development in the sapphire devil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Perchlorsäure, ACS reagent, 70%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%