Merck
  • Characterization of the sebocyte lipid droplet proteome reveals novel potential regulators of sebaceous lipogenesis.
Characterization of the sebocyte lipid droplet proteome reveals novel potential regulators of sebaceous lipogenesis.
Experimental cell research (2014-12-20)
Maik Dahlhoff, Thomas Fröhlich, Georg J Arnold, Udo Müller, Heinrich Leonhardt, Christos C Zouboulis, Marlon R Schneider
ZUSAMMENFASSUNG
Lipid metabolism depends on lipid droplets (LD), cytoplasmic structures surrounded by a protein-rich phospholipid monolayer. Although lipid synthesis is the hallmark of sebaceous gland cell differentiation, the LD-associated proteins of sebocytes have not been evaluated systematically. The LD fraction of SZ95 sebocytes was collected by density gradient centrifugation and associated proteins were analyzed by nanoliquid chromatography/tandem mass spectrometry. 54 proteins were significantly enriched in LD fractions, and 6 of them have not been detected previously in LDs. LD fractions contained high levels of typical LD-associated proteins as PLIN2/PLIN3, and most proteins belonged to functional categories characteristic for LD-associated proteins, indicating a reliable dataset. After confirming expression of transcripts encoding the six previously unidentified proteins by qRT-PCR in SZ95 sebocytes and in another sebocyte line (SebE6E7), we focused on two of these proteins, ALDH1A3 and EPHX4. While EPHX4 was localized almost exclusively on the surface of LDs, ALDH1A3 showed a more widespread localization that included additional cytoplasmic structures. siRNA-mediated downregulation revealed that depletion of EPHX4 increases LD size and sebaceous lipogenesis. Further studies on the roles of these proteins in sebocyte physiology and sebaceous lipogenesis may indicate novel strategies for the therapy of sebaceous gland-associated diseases such as acne.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Supelco
Natriumfluorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Linolsäure, analytical standard
Sigma-Aldrich
Natriumfluorid, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Natriumfluorid, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumfluorid 0,5 M -Lösung
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
Fluorid-Ionenlösung für ISE, 0.1 M F-, for ion-selective electrodes