Merck
Elatumic acid: a new ursolic acid congener from Omphalocarpum elatum Miers (Sapotaceae).
Zeitschrift fur Naturforschung. C, Journal of biosciences (2014-10-01)
Louis P Sandjo, Chi G Fru, Victor Kuete, Frederic Nana, Samuel O Yeboah, Renameditswe Mapitse, Berhanu M Abegaz, Thomas Efferth, Till Opatz, Bonaventure T Ngadjui
ZUSAMMENFASSUNG
A new triterpene diastereomer, 1, of the previously reported 3beta,6beta,19alpha-trihydroxy-urs-12-en-28-oic acid-24-carboxylic acid methyl ester was obtained from the stem bark of Omphalocarpum elatum Miers (Sapotaceae) along with a-amyrin acetate (2), spinasterol (3), spinasterol 3-O-beta-D-glucopyranoside (4), and tormentic acid (5). The structures of the isolates were established on the basis of NMR and mass spectrometric data and by comparison with those previously reported in the literature. Compound 1 showed weak antibacterial activity against E. aerogenes ATCC13048 and EA3, K. pneumoniae ATCC29916, and P aeruginosa; it also displayed moderate cytotoxicity against CCRF-CEM, CEM/ADR5000, and MDA-MB231 cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)