Merck
Altered gene expression in HepG2 cells exposed to a methanolic coal dust extract.
Environmental toxicology and pharmacology (2014-10-13)
Angelica Guerrero-Castilla, Jesus Olivero-Verbel
ZUSAMMENFASSUNG
Exposure to coal dust has been associated with different chronic diseases and mortality risk. This airborne pollutant is produced during coal mining and transport activities, generating environmental and human toxicity. The aim of this study was to determine the effects of a coal dust methanolic extract on HepG2, a human liver hepatocellular carcinoma cell line. Cells were exposed to 5-100ppm methanolic coal extract for 12h, using DMSO as control. MTT and comet assays were used for the evaluation of cytotoxicity and genotoxicity, respectively. Real time PCR was utilized to quantify relative expression of genes related to oxidative stress, xenobiotic metabolism and DNA damage. Coal extract concentrations did not induce significant changes in HepG2 cell viability after 12h exposure; however, 50 and 100ppm of the coal extract produced a significant increase in genetic damage index with respect to negative control. Compared to vehicle control, mRNA CYP1A1 (up to 163-fold), NQO1 (up to 4.7-fold), and GADD45B (up to 4.7-fold) were up regulated, whereas PRDX1, SOD, CAT, GPX1, XPA, ERCC1 and APEX1 remained unaltered. This expression profile suggests that cells exposed to coal dust extract shows aryl hydrocarbon receptor-mediated alterations, changes in cellular oxidative status, and genotoxic effects. These findings share some similarities with those observed in liver of mice captured near coal mining areas, and add evidence that living around these industrial operations may be negatively impacting the biota and human health.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture