Merck
Membrane activity of the pentaene macrolide didehydroroflamycoin in model lipid bilayers.
Biochimica et biophysica acta (2014-12-03)
Alena Koukalová, Šárka Pokorná, Radovan Fišer, Vladimír Kopecký, Jana Humpolíčková, Jan Černý, Martin Hof
ZUSAMMENFASSUNG
Didehydroroflamycoin (DDHR), a recently isolated member of the polyene macrolide family, was shown to have antibacterial and antifungal activity. However, its mechanism of action has not been investigated. Antibiotics from this family are amphiphilic; thus, they have membrane activity, their biological action is localized in the membrane, and the membrane composition and physical properties facilitate the recognition of a particular compound by the target organism. In this work, we use model lipid membranes comprised of giant unilamellar vesicles (GUVs) for a systematic study of the action of DDHR. In parallel, experiments are conducted using filipin III and amphotericin B, other members of the family, and the behavior observed for DDHR is described in the context of that of these two heavily studied compounds. The study shows that DDHR disrupts membranes via two different mechanisms and that the involvement of these mechanisms depends on the presence of cholesterol. The leakage assays performed in GUVs and the conductance measurements using black lipid membranes (BLM) reveal that the pores that develop in the absence of cholesterol are transient and their size is dependent on the DDHR concentration. In contrast, cholesterol promotes the formation of more defined structures that are temporally stable.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Decan, ReagentPlus®, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycero-3-Phosphocholin, lyophilized powder
Sigma-Aldrich
Decan, ≥95%
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Decan, anhydrous, ≥99%
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Supelco
Decan, analytical standard
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture