Merck
  • EGFR signaling downstream of EGF regulates migration, invasion, and MMP secretion of immortalized cells derived from human ameloblastoma.
EGFR signaling downstream of EGF regulates migration, invasion, and MMP secretion of immortalized cells derived from human ameloblastoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-08-08)
Marina Rolo Pinheiro da Rosa, Aline Semblano Carreira Falcão, Hellen Thais Fuzii, Maria Sueli da Silva Kataoka, André L R Ribeiro, Enrique Boccardo, Adriane Sousa de Siqueira, Ruy G Jaeger, João de Jesus Viana Pinheiro, Sérgio de Melo Alves Júnior
ZUSAMMENFASSUNG
Ameloblastoma is an odontogenic tumor characterized by local invasiveness and frequent recurrence. The surrounding stroma, composed of different cell types and extracellular matrix (ECM), may influence ameloblastoma invasive behavior. Furthermore, tumor and stromal cells secrete matrix metalloproteases (MMPs), which, in turn, can modulate the matrix and promote the release of ECM-bound growth factors. Among these growth factors, epidermal growth factor (EGF) and its receptor, EGFR, have already been shown to stimulate MMP synthesis, suggesting that an interdependent mechanism, involving MMP activity and growth factors release, may contribute to tumor invasiveness. The aim of this study was to evaluate the effects of the EGF/EGFR signaling pathway on migration, invasion, and MMP activity, in a primary cell line derived from human ameloblastoma. We established and characterized a primary cell line (AME-1) from a human ameloblastoma sample. This cell line was transduced with human papillomavirus type 16 (HPV16) E6/E7 oncogenes, generating the AME-HPV continuous cell line. EGF, MMP2, and MMP9 expression in ameloblastoma biopsies and in the AME-HPV cell line was analyzed by immunohistochemistry and immunofluorescence, respectively. Migratory activity of EGF-treated AME-HPV cells was investigated using monolayer wound assays and Transwell chambers. EGF-induced invasion was assessed in Boyden chambers coated with Matrigel. Conditioned medium from EGF-treated cells was subjected to zymography. EGFR expression in AME-HPV cells was silenced by small interfering RNA (siRNA), to verify the relationship between this receptor and MMP secretion. Ameloblastoma samples and AME-HPV cells expressed EGF, EGFR, MMP2, and MMP9. AME-HPV cells treated with EGF showed increased rates of migration and invasion, as well as enhanced MMP2 and MMP9 activity. EGFR knockdown decreased MMP2 and MMP9 levels in AME-HPV cells. EGFR signaling downstream of EGF probably regulates migration, invasion, and MMP secretion of ameloblastoma-derived cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hämatoxylin
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)