Merck
  • GC-MS analysis of organic acids in human urine in clinical settings: a study of derivatization and other analytical parameters.
GC-MS analysis of organic acids in human urine in clinical settings: a study of derivatization and other analytical parameters.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-02-01)
Chrysoula Christou, Helen G Gika, Nikolaos Raikos, Georgios Theodoridis
ZUSAMMENFASSUNG
In the current paper the analytical conditions for the determination of ten free organic acids by GC-MS are studied with the aim to establish a method for organic acid profiling in human urine to be used as a tool for the detection of metabolic or other health disorders. Studies included the GC-MS method development, the derivatization (trimethylsilylation) reaction conditions, the stability of the standard solutions during storage in the freezer, and the stability of the formed trimethylsilyl derivatives. Best results were obtained at a derivatization temperature of 50°C, and a reaction time of 30 min. Standard solutions were stable for 22 days, derivatized samples were stable at least for 30 h when stored at -24°C. GC-MS analysis achieved sensitive determination of the organic acids under study with limits of detection ranging from 0.03 mmol/mol creatinine for glutaric acid, to 0.34 mmol/mol creatinine for glycolic acid. Within-day and day-to-day assay imprecision was found satisfactory with relative standard deviations being below 10%. The developed method was successfully applied to the quantitative analysis of free organic acids in urine samples obtained from hospitalized children. Creatinine-corrected excretion rates of all analyzed organic acids were within reference intervals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, derivatization grade (GC derivatization), LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Chlorotrimethylsilan, ≥98.0% (GC)
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Natriumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Adipinsäure, 99%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoracetamid, ≥99%