Merck
  • Epidemiology and identification of potential fungal pathogens causing invasive fungal infections in a tertiary care hospital in northeast Thailand.
Epidemiology and identification of potential fungal pathogens causing invasive fungal infections in a tertiary care hospital in northeast Thailand.
Medical mycology (2014-09-19)
Kiatichai Faksri, Wanlop Kaewkes, Kunyaluk Chaicumpar, Prajuab Chaimanee, Suwin Wongwajana
ZUSAMMENFASSUNG
Invasive fungal infections (IFIs) are life threatening and associated with a high mortality rate. Here, we describe the distribution of pathogens, host risk factors, and significance of fungi isolated from patients with IFIs. The study included 861 fungal isolates recovered between 2006 and 2011 from 802 patients at Srinagarind Hospital, Thailand. Based on the European Organization for Research and Treatment of Cancer/Invasive Fungal Infections Cooperative Group and the National Institute of Allergy and Infectious Diseases Mycoses Study Group 2008 criteria, 28.5% (245/861 isolates) of the fungal isolates were considered to be causative agents of IFIs. The most common fungus was Candida albicans (46%, 396/861 isolates). However, the most common yeast causing IFIs was Cryptococcus neoformans (34.7%, 85/245 isolates), while the most common mould was Penicillium marneffei (10.6%, 26/245 isolates). Cryptococcosis was significantly associated with human immunodeficiency virus infections (P < 0.001). Trend analysis revealed that there was no significant increase in IFI cases (P = 0.34) from 2006 to 2011 or from 2007 to 2011 (P = 0.05), but there was a trend toward significant increases in candidiasis (P = 0.04). The fungal isolates were categorized according to the positive predictive value of their recovery in cultures as being true (>95%), moderate (5%-95%), and rare (<5%) pathogens. This classification system could facilitate the prediction of the likelihood of diseases caused by the isolated fungi.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard