Merck
WS0701: a novel sedative-hypnotic agent acting on the adenosine system.
Behavioural pharmacology (2014-08-30)
Xiao-Yu Bai, Xue-Qiong Zhang, Yong-He Zhang, Song Wu, Ling-Hua Hao, Rui Liu, Zhong-Lin Huang, Wei-Ku Zhang, Zong-Miao Sun, Guan-Hua Du
ZUSAMMENFASSUNG
To characterize the sedative and hypnotic profile of the novel adenosine derivative ((3S,4R,5R)-3,4-dihydroxy-5-(6-((4-hydroxy-3-methoxybenzyl)amino)-9H-purin-9-yl)tetrahydrofuran-2-yl) methyl diaconate (WS0701), we performed a variety of behavioural tests and investigated the influence of WS0701 on various sleep stages. In mice, WS0701 significantly increased the number of entries and time spent in open arms in the elevated plus maze test, indicating an anxiolytic effect. WS0701 decreased locomotor activity counts and head dips in the hole-board test and enhanced sodium pentobarbital-induced hypnosis. However, WS0701 did not induce the loss of the righting reflex or amnesic effects in behavioural models. In rats, WS0701 exerted a sedative effect and markedly prolonged the time spent in non-rapid-eye-movement sleep, especially slow-wave sleep, but reduced the time spent in rapid-eye-movement sleep (REMS). Pretreatment with the selective adenosine A2a receptor antagonist SCH58261 attenuated the sedative and hypnotic effects of WS0701. WS0701 did not protect mice against picrotoxin-induced seizures, but inhibited adenosine deaminase activity and increased adenosine levels in the frontal cortex and hypothalamus of mice. In conclusion, WS0701 shows anxiolytic, sedative as well as sleep stage alterative effects, which may be related to the adenosine system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Adenosin, ≥99%
Sigma-Aldrich
Hypoxanthin, ≥99.0%
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Sigma-Aldrich
Hypoxanthin, powder, BioReagent, suitable for cell culture
USP
Koffein, United States Pharmacopeia (USP) Reference Standard
Supelco
Schmelzpunktstandard 235-237°C, analytical standard
Supelco
Coffein Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adenosin, BioReagent, suitable for cell culture
Supelco
Koffein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Koffein-Schmelzpunkt-Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Koffein, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Koffein, meets USP testing specifications, anhydrous
Sigma-Aldrich
Adenosin
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Supelco
Mettler Toledo Calibration Substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Koffein, anhydrous, tested according to Ph. Eur.
Supelco
Adenosin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Koffein, BioXtra
Koffein für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Koffein, European Pharmacopoeia (EP) Reference Standard
Adenosin, European Pharmacopoeia (EP) Reference Standard
Didanosin Unreinheit A,, European Pharmacopoeia (EP) Reference Standard