Merck
  • Promising potential of new generation translocator protein tracers providing enhanced contrast of arthritis imaging by positron emission tomography in a rat model of arthritis.
Promising potential of new generation translocator protein tracers providing enhanced contrast of arthritis imaging by positron emission tomography in a rat model of arthritis.
Arthritis research & therapy (2014-03-15)
Yoony Y J Gent, Karin Weijers, Carla F M Molthoff, Albert D Windhorst, Marc C Huisman, Michael Kassiou, Gerrit Jansen, Adriaan A Lammertsma, Conny J van der Laken
ZUSAMMENFASSUNG
Early diagnosis of and subsequent monitoring of therapy for rheumatoid arthritis (RA) could benefit from detection of (sub)clinical synovitis. Imaging of (sub)clinical arthritis by targeting the translocator protein (TSPO) on activated macrophages is feasible using (R)-[¹¹C] PK11195-based positron emission tomography (PET), but clinical applications are limited by background uptake in peri-articular bone/bone marrow. The purpose of the present study was to evaluate two other TSPO ligands with potentially lower background uptake in neurological studies, [¹¹C]DPA-713 and [¹⁸F]DPA-714, in a rat model of arthritis. TSPO binding of DPA-713, DPA-714 and PK11195 were assessed by in vitro competition studies with [³H]DPA-713 using human macrophage THP-1 cells and CD14⁺ monocytes from healthy volunteers. In vivo studies were performed in rats with methylated bovine serum albumin-induced knee arthritis. Immunohistochemistry with anti-TSPO antibody was performed on paraffin-embedded sections. Rats were imaged with [¹¹C]DPA-713 or [¹⁸F]DPA-714 PET, followed by ex vivo tissue distribution studies. Results were compared with those obtained with the tracer (R)-[¹¹C]PK11195, the established ligand for TSPO. In THP-1 cells, relative TSPO binding of DPA-713 and DPA-714 were 7-fold and 25-fold higher, respectively, than in PK11195. Comparable results were observed in CD14⁺ monocytes from healthy volunteers. In the arthritis rat model, immunohistochemistry confirmed the presence of TSPO-positive inflammatory cells in the arthritic knee. PET images showed that uptake of [¹¹C]DPA-713 and [¹⁸F]DPA-714 in arthritic knees was significantly increased compared with contralateral knees and knees of normal rats. Uptake in arthritic knees could be largely blocked by an excess of PK11195. [¹¹C]DPA-713 and [¹⁸F]DPA-714 provided improved contrast compared with (R)-[¹¹C]PK11195, as was shown by significantly higher arthritic knee-to-bone ratios of [¹¹C]DPA-713 (1.60 ± 0.31) and [¹⁸F]DPA-714 (1.55 ± 0.10) compared with (R)-[¹¹C]PK11195 (1.14 ± 0.19). [¹¹C]DPA-713 and [¹⁸F]DPA-714 clearly visualized arthritis and exhibited lower (peri-articular) bone/bone marrow uptake than (R)-[¹¹C]PK11195. These features merit further investigation of these tracers for early diagnosis and therapy monitoring of RA in a clinical setting.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture