Merck
  • Interleukin-6 and interleukin-10 gene promoter polymorphisms and risk of endemic Burkitt lymphoma.
Interleukin-6 and interleukin-10 gene promoter polymorphisms and risk of endemic Burkitt lymphoma.
The American journal of tropical medicine and hygiene (2014-07-30)
Cliff I Oduor, Kiprotich Chelimo, Collins Ouma, David H Mulama, Joslyn Foley, John Vulule, Jeffrey A Bailey, Ann M Moormann
ZUSAMMENFASSUNG
Overexpression of interleukin-6 (IL-6) and IL-10 in endemic Burkitt lymphoma (eBL) may facilitate tumorigenesis by providing a permissive cytokine milieu. Promoter polymorphisms influence interindividual differences in cytokine production. We hypothesized that children genetically predisposed for elevated cytokine levels may be more susceptible to eBL. Using case-control samples from western Kenya consisting of 117 eBL cases and 88 ethnically matched healthy controls, we tested for the association between eBL risk and IL-10 (rs1800896, rs1800871, and rs1800872) and IL-6 (rs1800795) promoter single nucleotide polymorphisms (SNPs) as well as IL-10 promoter haplotypes. In addition, the association between these variants and Epstein Barr Virus (EBV) load was examined. Results showed that selected IL-10 and IL-6 promoter SNPs and IL-10 promoter haplotypes were not associated with risk eBL or EBV levels in EBV-seropositive children. Findings from this study reveal that common variants within the IL-10 and IL-6 promoters do not independently increase eBL risk in this vulnerable population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)