Merck
  • Expression of syntaxin 8 in visceral adipose tissue is increased in obese patients with type 2 diabetes and related to markers of insulin resistance and inflammation.
Expression of syntaxin 8 in visceral adipose tissue is increased in obese patients with type 2 diabetes and related to markers of insulin resistance and inflammation.
Archives of medical research (2014-12-20)
Andoni Lancha, Santiago López-Garrido, Amaia Rodríguez, Victoria Catalán, Beatriz Ramírez, Víctor Valentí, Rafael Moncada, Camilo Silva, María J Gil, Javier Salvador, Gema Frühbeck, Javier Gómez-Ambrosi
ZUSAMMENFASSUNG
Obesity is associated with increased adipose tissue inflammation as well as with the development of type 2 diabetes (T2D). Syntaxin 8 (STX8) is a protein required for the transport of endosomes. In this study we analyzed the relationship of STX8 with the presence of T2D in the context of obesity. With this purpose, 21 subjects (seven lean [LN], eight obese normoglycemic [OB-NG] and six obese with type 2 diabetes [OB-T2D]) were included in the study. Gene and protein expression levels of STX8 and GLUT4 were analyzed in visceral adipose tissue (VAT). mRNA (p = 0.008) and protein (p <0.001) expression levels of STX8 were significantly increased in VAT of OB-T2D patients. Moreover, gene expression levels of SLC2A4 (GLUT4) were downregulated (p = 0.002) in VAT of obese patients. We found that STX8 was positively correlated (p <0.05) with fasting glucose concentrations, plasma glucose 2 h after an OGTT and C-reactive protein. Interestingly, the expression of STX8 was negatively correlated (p <0.05) with the expression of SLC2A4 in VAT. Increased STX8 expression in VAT appears to be associated with the presence of T2D in obese patients through a mechanism that may involve GLUT4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri