Merck
  • Genome annotation by shotgun inactivation of a native gene in hemizygous cells: application to BRCA2 with implication of hypomorphic variants.
Genome annotation by shotgun inactivation of a native gene in hemizygous cells: application to BRCA2 with implication of hypomorphic variants.
Human mutation (2014-12-03)
Soma Ghosh, Anil K Bhunia, Bogdan C Paun, Samuel F Gilbert, Urmil Dhru, Kalpesh Patel, Scott E Kern
ZUSAMMENFASSUNG
The greatest interpretive challenge of modern medicine may be to functionally annotate the vast variation of human genomes. Demonstrating a proposed approach, we created a library of BRCA2 exon 27 shotgun-mutant plasmids including solitary and multiplex mutations to generate human knockin clones using homologous recombination. This 55-mutation, 13-clone syngeneic variance library (SyVaL) comprised severely affected clones having early-stop nonsense mutations, functionally hypomorphic clones having multiple missense mutations emphasizing the potential to identify and assess hypomorphic mutations in novel proteomic and epidemiologic studies, and neutral clones having multiple missense mutations. Efficient coverage of nonessential amino acids was provided by mutation multiplexing. Severe mutations were distinguished from hypomorphic or neutral changes by chemosensitivity assays (hypersensitivity to mitomycin C and acetaldehyde), by analysis of RAD51 focus formation, and by mitotic multipolarity. A multiplex unbiased approach of generating all-human SyVaLs in medically important genes, with random mutations in native genes, would provide databases of variants that could be functionally annotated without concerns arising from exogenous cDNA constructs or interspecies interactions, as a basis for subsequent proteomic domain mapping or clinical calibration if desired. Such gene-irrelevant approaches could be scaled up for multiple genes of clinical interest, providing distributable cellular libraries linked to public-shared functional databases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acetaldehyd, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyd, natural, FG
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyd, puriss. p.a., anhydrous, ≥99.5% (GC)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, contains NaCl as solubilizer
Supelco
Acetaldehyd, PESTANAL®, analytical standard