Merck
Relating toxicity to transfection: using sphingosine to maintain prolonged expression in vitro.
Molecular pharmaceutics (2014-11-25)
Jamie L Betker, Thomas J Anchordoquy
ZUSAMMENFASSUNG
Cationic reagents are commonly used to facilitate DNA delivery, and transfection experiments are typically initiated in cell culture where the optimal charge ratio is determined. While transfection rates are often enhanced at higher +/- charge ratios, the cellular toxicity associated with the greater amounts of cationic components at elevated charge ratios is often not considered. In addition, the prolonged effects of cationic lipid uptake on cell viability are not evident in a typical 24-48 h transfection experiment. In this study, we compare the transfection efficiency of cationic lipoplexes to effects on viability of cultured cells in both the short and long term (7 days). Our results indicate that, while minimal toxicity is evident 24 h after exposure to DOTAP-based lipoplexes, cell viability continues to decline and ultimately compromises reporter gene expression at longer times. Substitution of a naturally occurring cationic amphiphile, sphingosine, for DOTAP greatly reduces toxicity and allows high expression to be maintained over prolonged periods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
D-Sphingosin, synthetic
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.