Merck
  • Polydopamine-based immobilization of zeolitic imidazolate framework-8 for in-tube solid-phase microextraction.
Polydopamine-based immobilization of zeolitic imidazolate framework-8 for in-tube solid-phase microextraction.
Journal of chromatography. A (2015-03-03)
Juan Zhang, Wenpeng Zhang, Tao Bao, Zilin Chen
ZUSAMMENFASSUNG
Zeolitic imidazolate frameworks (ZIFs), a subfamily of metal-organic frameworks (MOFs), have received considerable attention as a novel porous material for sample preparation due to their fascinating structures and unique properties. Here, we developed a novel bio-inspired polydopamine (PDA) method to immobilize ZIFs inside the chemical resistance poly(ether ether ketone) (PEEK) tube for online solid-phase microextraction (SPME). Firstly, PDA layer was assembled inside the PEEK tube. Secondly, attributing to its noncovalent adsorption and covalent reaction ability, PDA could attract and anchor ZIF-8 nutrients onto the inner surface of PEEK tube to promote the nucleation and growth of ZIF-8. The morphology and surface properties of the prepared ZIF-8-PDA-PEEK tube were characterized by scanning electron microscopy and energy dispersive X-ray spectrometry. The ZIF-8-PDA-PEEK tube exhibits excellent extraction efficiency toward six polycyclic aromatic hydrocarbons (PAHs), with enrichment factor from 550 to 734. The developed online SPME-HPLC method shows good linearity (10-5000pg/mL) and low detection limits (0.5-5 pg/mL) for six PAHs. It also has been used to determine PAHs in environmental samples, with recoveries in the range of 82.5-98.6%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dopamin -hydrochlorid
Sigma-Aldrich
Benzo[a]pyren, ≥96% (HPLC)
Sigma-Aldrich
Pyren, 98%
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
Benzo[a]pyren, analytical standard, for environmental analysis
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
Benzo[a]pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Sigma-Aldrich
Pyren, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
Pyren, analytical standard
Supelco
Dopamin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benz[a]anthracen, 99%
Sigma-Aldrich
Fluoranthen, 98%
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS/DVB
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Fluoranthen, analytical standard
Supelco
Benz[a]anthracen, analytical standard
Supelco
SPME-Faseranordnung Polydimethylsiloxan/Divinylbenzen (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME-Faseranordnung Polyacrylat (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
Tragbarer SPME-Field-Sampler, coating PDMS
Supelco
SPME StableFlex Methodenentwicklungs-Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME StableFlex Methodenentwicklungs-Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4