Merck
  • Diarrhoea caused by rotavirus in a regional Peruvian hospital: determination of circulating genotypes.
Diarrhoea caused by rotavirus in a regional Peruvian hospital: determination of circulating genotypes.
Transactions of the Royal Society of Tropical Medicine and Hygiene (2014-04-30)
Pablo Weilg Espejo, Fiorella Orellana Peralta, Hernán Cornejo Pacheres, Luis J del Valle, Angela Cornejo Tapia, Jorge Bazán Mayra, Joaquim Ruiz, Juana Del Valle Mendoza
ZUSAMMENFASSUNG
Gastroenteritis caused by rotavirus is responsible for approximately 810 deaths per year in children under 5 years of age in Peru and emerging rotavirus genotypes have led to concerns regarding cross-protection by the vaccines available. Moreover, there are no reports on the molecular epidemiology of rotavirus diarrhoea in Peru. A total of 131 stool samples were obtained from children under 5 years hospitalised from January 2010 to December 2012 in the Hospital Regional de Cajamarca (Peru). ELISA and RT-PCR techniques were performed for rotavirus detection. G and P typing of rotavirus-positive samples were performed by semi-nested multiplex RT-PCR, and sequencing was performed to confirm the PCR results. Of the 117 samples available, 22 (18.8%) tested positive for rotavirus by ELISA and 42 (35.9%) tested positive by RT-PCR. Among the G genotypes identified, G9 (35.7%; 15/42) and G12 (33.3%; 14/42) were the most prevalent, with the most common combination being G12/P[6] (23.8%; 10/42). A high prevalence of the G12/P[6] genotype was detected. It is known that this genotype is not covered by the current vaccines available. More in-depth studies are needed to determine the current rotavirus genotypes presents in Peru.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O