Merck
Heterogeneous nucleation and shape transformation of multicomponent metallic nanostructures.
Nature materials (2014-11-05)
Soon Gu Kwon, Galyna Krylova, Patrick J Phillips, Robert F Klie, Soma Chattopadhyay, Tomohiro Shibata, Emilio E Bunel, Yuzi Liu, Vitali B Prakapenka, Byeongdu Lee, Elena V Shevchenko
ZUSAMMENFASSUNG
To be able to control the functions of engineered multicomponent nanomaterials, a detailed understanding of heterogeneous nucleation at the nanoscale is essential. Here, by using in situ synchrotron X-ray scattering, we show that in the heterogeneous nucleation and growth of Au on Pt or Pt-alloy seeds the heteroepitaxial growth of the Au shell exerts high stress (∼2 GPa) on the seed by forming a core/shell structure in the early stage of the reaction. The development of lattice strain and subsequent strain relaxation, which we show using atomic-resolution transmission electron microscopy to occur through the slip of {111} layers, induces morphological changes from a core/shell to a dumbbell structure, and governs the nucleation and growth kinetics. We also propose a thermodynamic model for the nucleation and growth of dumbbell metallic heteronanostructures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Didodecyldimethylammoniumbromid, 98%
Sigma-Aldrich
Hexadecylamin, 98%
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG
Sigma-Aldrich
Hexadecylamin, technical grade, 90%
Supelco
Citral, analytical standard