Merck
  • Peripheral inflammation activated focal adhesion kinase signaling in spinal dorsal horn of mice.
Peripheral inflammation activated focal adhesion kinase signaling in spinal dorsal horn of mice.
Journal of neuroscience research (2015-01-24)
Xia Lian, Xin-Tai Wang, Wen-Tao Wang, Xian Yang, Zhan-Wei Suo, Xiao-Dong Hu
ZUSAMMENFASSUNG
Focal adhesion kinase (FAK) is one of the nonreceptor protein tyrosine kinases critical for the dynamic regulation of cell adhesion structures. Recent studies have demonstrated that FAK is also localized at excitatory glutamatergic synapses and is involved in long-term modification of synaptic strength. However, whether FAK is engaged in nociceptive processing in the spinal dorsal horn remains unresolved. The current study shows that intraplantar injection of complete Freund's adjuvant (CFA) in mice significantly increases FAK autophosphorylation at Tyr397, indicating a close correlation of FAK activation with inflammatory pain. FAK activation depended on the activity of N-methyl-D-aspartate-subtype glutamate receptor (NMDAR) and metabotropic glutamate receptor (mGluR) because pharmacological inhibition of NMDAR or group I mGluR totally abolished FAK phosphorylation induced by CFA. The active FAK operated to stimulate extracellular signal-regulated kinase1/2 (ERK1/2), which boosted the protein expression of GluN2B subunit-containing NMDAR at the synaptosomal membrane fraction. Inhibition of FAK activity by spinal expression of a kinase-dead FAK(Y397F) mutant repressed ERK1/2 hyperactivity and reduced the synaptic concentration of NMDAR in CFA-injected mice. Electrophysiological recording demonstrated that intracellular loading of specific anti-FAK antibody significantly reduced the amplitudes of NMDAR-mediated excitatory postsynaptic currents on lamina II neurons from inflamed mice but not from naive mice. Behavioral tests showed that spinal expression of FAK(Y397F) generated a long-lasting alleviation of CFA-induced mechanical allodynia and thermal hyperalgesia. These data indicate that FAK might exaggerate NMDAR-mediated synaptic transmission in the spinal dorsal horn to sensitize nociceptive behaviors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Millipore
Saccharose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF