Merck
  • LY293558 prevents soman-induced pathophysiological alterations in the basolateral amygdala and the development of anxiety.
LY293558 prevents soman-induced pathophysiological alterations in the basolateral amygdala and the development of anxiety.
Neuropharmacology (2014-09-11)
Eric M Prager, Taiza H Figueiredo, Robert P Long, Vassiliki Aroniadou-Anderjaska, James P Apland, Maria F M Braga
ZUSAMMENFASSUNG
Exposure to nerve agents can cause brain damage due to prolonged seizure activity, producing long-term behavioral deficits. We have previously shown that LY293558, a GluK1/AMPA receptor antagonist, is a very effective anticonvulsant and neuroprotectant against nerve agent exposure. In the present study, we examined whether the protection against nerve agent-induced seizures and neuropathology conferred by LY293558 translates into protection against pathophysiological alterations in the basolateral amygdala (BLA) and the development of anxiety, which is the most prevalent behavioral deficit resulting from exposure. LY293558 (15 mg/kg) was administered to rats, along with atropine and HI-6, at 20 min after exposure to soman (1.2 × LD50). At 24 h, 7 days, and 30 days after exposure, soman-exposed rats who did not receive LY293558 had reduced but prolonged evoked field potentials in the BLA, as well as increased paired-pulse ratio, suggesting neuronal damage and impaired synaptic inhibition; rats who received LY293558 did not differ from controls in these parameters. Long-term potentiation of synaptic transmission was impaired at 7 days after exposure in the soman-exposed rats who did not receive anticonvulsant treatment, but not in the LY293558-treated rats. Anxiety-like behavior assessed by the open field and acoustic startle response tests was increased in the soman-exposed rats at 30 and 90 days after exposure, while rats treated with LY293558 did not differ from controls. Along with our previous findings, the present data demonstrate the remarkable efficacy of LY293558 in counteracting nerve agent-induced seizures, neuropathology, pathophysiological alterations in the BLA, and anxiety-related behavioral deficits.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Atropin -sulfat (Salz) Monohydrat, ≥97% (TLC), crystalline
Supelco
Atropinsulfat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
USP
Atropinsulfat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Atropinsulfat, European Pharmacopoeia (EP) Reference Standard