Merck
  • Evans blue staining reveals vascular leakage associated with focal areas of host-parasite interaction in brains of pigs infected with Taenia solium.
Evans blue staining reveals vascular leakage associated with focal areas of host-parasite interaction in brains of pigs infected with Taenia solium.
PloS one (2014-06-11)
Miguel Marzal, Cristina Guerra-Giraldez, Adriana Paredes, Carla Cangalaya, Andrea Rivera, Armando E Gonzalez, Siddhartha Mahanty, Hector H Garcia, Theodore E Nash
ZUSAMMENFASSUNG
Cysticidal drug treatment of viable Taenia solium brain parenchymal cysts leads to an acute pericystic host inflammatory response and blood brain barrier breakdown (BBB), commonly resulting in seizures. Naturally infected pigs, untreated or treated one time with praziquantel were sacrificed at 48 hr and 120 hr following the injection of Evans blue (EB) to assess the effect of treatment on larval parasites and surrounding tissue. Examination of harvested non encapsulated muscle cysts unexpectedly revealed one or more small, focal round region(s) of Evans blue dye infiltration (REBI) on the surface of otherwise non dye-stained muscle cysts. Histopathological analysis of REBI revealed focal areas of eosinophil-rich inflammatory infiltrates that migrated from the capsule into the tegument and internal structures of the parasite. In addition some encapsulated brain cysts, in which the presence of REBI could not be directly assessed, showed histopathology identical to that of the REBI. Muscle cysts with REBI were more frequent in pigs that had received praziquantel (6.6% of 3736 cysts; n = 6 pigs) than in those that were untreated (0.2% of 3172 cysts; n = 2 pigs). Similar results were found in the brain, where 20.7% of 29 cysts showed histopathology identical to muscle REBI cysts in praziquantel-treated pigs compared to the 4.3% of 47 cysts in untreated pigs. Closer examination of REBI infiltrates showed that EB was taken up only by eosinophils, a major component of the cellular infiltrates, which likely explains persistence of EB in the REBI. REBI likely represent early damaging host responses to T. solium cysts and highlight the focal nature of this initial host response and the importance of eosinophils at sites of host-parasite interaction. These findings suggest new avenues for immunomodulation to reduce inflammatory side effects of anthelmintic therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar