Merck
  • Treatment with low-temperature atmospheric pressure plasma enhances cutaneous delivery of epidermal growth factor by regulating E-cadherin-mediated cell junctions.
Treatment with low-temperature atmospheric pressure plasma enhances cutaneous delivery of epidermal growth factor by regulating E-cadherin-mediated cell junctions.
Archives of dermatological research (2014-04-15)
Jeong-Hae Choi, Seoul-Hee Nam, Yeon-Suk Song, Hyun-Wook Lee, Hae-June Lee, Kiwon Song, Jin-Woo Hong, Gyoo-Cheon Kim
ZUSAMMENFASSUNG
The barrier system of the skin not only defends against antigens and harmful substances, but also hinders the permeation of medicines and cosmetics into the dermis. Several strategies have been developed to enhance the absorption ability of skin, including the use of chemicals and skin ablation devices. However, the cost and inconvenience of these strategies highlights the need for a novel and safe method for increasing skin absorption. In this study, we examined the effect of low temperature atmospheric pressure plasma (LTAPP) on the efficiency of drug penetration through the skin, as well as its mechanism of action. HaCaT human keratinocytes and hairless mice were exposed to LTAPP treatment, and the cellular and tissue gene expression, and morphological changes were monitored. We found that the LTAPP exposure reduced the expression of E-cadherin in skin cells and led to the loss of cell-cell contacts. The exposure of mouse skin to LTAPP also reduced the expression of E-cadherin and prevented intercellular junction formation within the tissue, leading to enhanced absorption of hydrophilic agents, eosin and epidermal growth factor. The reduction in E-cadherin expression and reduced skin barrier function recovered completely within 3 h of LTAPP exposure. Taken together, these data show that LTAPP can induce a temporal decrease in the skin barrier function by regulating E-cadherin-mediated intercellular interactions, leading to the enhanced transdermal delivery of drugs and cosmetics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eosin Y -Lösung, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y Dinatriumsalz, certified by the BSC
Sigma-Aldrich
Eosin Y Dinatriumsalz, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y, 75% (HPLC)