Merck
  • Intrinsic enzyme mimicking activity of gold nanoclusters upon visible light triggering and its application for colorimetric trypsin detection.
Intrinsic enzyme mimicking activity of gold nanoclusters upon visible light triggering and its application for colorimetric trypsin detection.
Biosensors & bioelectronics (2014-10-14)
Guang-Li Wang, Lu-Yi Jin, Yu-Ming Dong, Xiu-Ming Wu, Zai-Jun Li
ZUSAMMENFASSUNG
In this research, a novel enzyme mimetics based on the photochemical property of gold nanoclusters was demonstrated. It was found that the bovine serum albumin (BSA) stabilized red or blue emitting gold nanoclusters (Au NCs) exhibited enzyme-like activity under visible light irradiation. The BSA-Au NCs had better stability against stringent conditions compared to natural enzyme. In addition, the photostimulated enzyme mimetics of BSA-Au NCs showed several unprecedented advantages over natural peroxidase or other existing alternatives based on nanomaterials, such as the independence of hydrogen peroxide on activity and the easily regulated activity by light irradiation. The mechanism of the photoresponsive enzyme-like activity of BSA-Au NCs was investigated. The photoactivated BSA-Au NCs was designed to develop a facile, cheap, and rapid colorimetric assay to detect trypsin through trypsin digestion of the protein template of BSA-stabilized Au NCs. The limit of detection for trypsin was 0.6 μg/mL, which was much lower than the average level of trypsin in patient's urine or serum.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
tert-Butanol, analytical standard