Merck
  • Characterization of bacteria degrading 3-hydroxy palmitic acid methyl ester (3OH-PAME), a quorum sensing molecule of Ralstonia solanacearum.
Characterization of bacteria degrading 3-hydroxy palmitic acid methyl ester (3OH-PAME), a quorum sensing molecule of Ralstonia solanacearum.
Letters in applied microbiology (2015-01-13)
G A Achari, R Ramesh
ZUSAMMENFASSUNG
Bacterial wilt pathogen Ralstonia solanacearum causes severe crop loss of eggplant, which is of economic importance in India. 3-hydroxy palmitic acid methyl ester (3OH-PAME) is the main quorum sensing molecule governing the expression of virulence factors in R. solanacearum. Ability of 164 bacterial isolates from the xylem of eggplant (Solanum melongena L.), chilli pepper (Capsicum annuum L.) and wild eggplant (Solanum torvum Sw.) to degrade 3OH-PAME was tested by disc diffusion assay. Enzymatic degradation of 3OH-PAME by five bacteria was confirmed by High-Performance Liquid Chromatography-Mass Spectrometry analysis. 3OH-PAME degrading bacteria were identified as Stenotrophomonas maltophilia, Pseudomonas aeruginosa and Rhodococcus corynebacterioides. 3OH-PAME degrading bacteria reduced the expression of virulence factors (exopolysaccharides and endoglucanase) of R. solanacearum in vitro and reduced wilt incidence in eggplant seedlings under greenhouse conditions. Isolates with quorum quenching activity successfully re-colonized eggplant seedlings. Quorum quenching bacteria produced antagonistic compounds, which may act synergistically with quorum quenching in reducing bacterial wilt in eggplant. This is the first report on endophytic bacteria of class Gammaproteobacteria and phylum Actinobacteria having 3OH-PAME degrading activity. This study demonstrates the potential use of endophytic bacteria as quorum quenching biocontrol agents for management of bacterial wilt in eggplant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Supelco
Dichlormethan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Nalidixinsäure, ≥98%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Dichlormethan, analytical standard
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Chlorwasserstoff–2-Propanol -Lösung, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Nalidixinsäure, analytical standard