Merck
Nitidine chloride inhibits hepatic cancer growth via modulation of multiple signaling pathways.
BMC cancer (2014-10-01)
Jiumao Lin, Aling Shen, Hongwei Chen, Jun Liao, Teng Xu, Liya Liu, Jing Lin, Jun Peng
ZUSAMMENFASSUNG
The development of hepatic cancer is tightly regulated by multiple intracellular signaling pathways. Therefore, most currently-used anti-tumor agents, which typically target single intracellular pathway, might not always be therapeutically effective. Additionally, long-term use of these agents probably generates drug resistance and unacceptable adverse effects. These problems increase the necessity for the development of new chemotherapeutic approaches. Nitidine chloride (NC), a natural benzophenanthridine alkaloid, has been shown to inhibit cancer growth via induction of cell apoptosis and suppression of cancer angiogenesis. But the precise mechanisms of its tumorcidal activity are not well understood. To further elucidate the precise mechanisms of its anti-tumor activity, using a hepatic cancer mouse xenograft model, the human hepatic cancer cell lines (HepG2, HCCLM3, Huh7), and umbilical vein endothelial cells (HUVEC), here we evaluate the effect of NC on tumor growth in vivo and in vitro and investigated the underlying molecular mechanisms. We found that NC treatment resulted in significant decrease in tumor volume and tumor weight respectively, but didn't affect body weight changes. Additionally, NC treatment dose- and time-dependently reduced the cell viability of all three hepatic cell lines. Moreover, NC suppressed the activation of STAT3, ERK and SHH pathways; and altered the expression of critical target genes including Bcl-2, Bax, Cyclin D1, CDK4, VEGF-A and VEGFR2. These molecular effects resulted in the promotion of apoptosis, inhibition of cell proliferation and tumor angiogenesis. Our findings suggest that NC possesses a broad range of anti-cancer activities due to its ability to affect multiple intracellular targets, suggesting that NC could be a novel multi-potent therapeutic agent for the treatment of hepatic cancer and other cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Benzimidazol, 98%
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar