Merck
  • Carboxymethylcellulose-tetrahydrocurcumin conjugates for colon-specific delivery of a novel anti-cancer agent, 4-amino tetrahydrocurcumin.
Carboxymethylcellulose-tetrahydrocurcumin conjugates for colon-specific delivery of a novel anti-cancer agent, 4-amino tetrahydrocurcumin.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-05-27)
Thipapun Plyduang, Luelak Lomlim, Supreeya Yuenyongsawad, Ruedeekorn Wiwattanapatapee
ZUSAMMENFASSUNG
Several curcumin derivatives are now becoming increasingly of interest because of their bioactive attributes, especially their action as antioxidants and anti-carcinogenic activities. Tetrahydrocurcumin (THC), an active metabolite of curcumin, was selected to be a proper starting material for the work presented here as it is stable in physiological pH and has the typical pharmacological properties of curcumin. We have now reported that novel synthesized water-soluble polymeric macromolecule prodrugs can specifically deliver the drug to the colon. To study the drug loading and drug release, THC was conjugated with a hydrophilic polymer, carboxymethylcellulose (CMC) with the degree of substitution (DS) values of 0.7 and 1.2. THC was also attached to two different spacers including p-aminobenzoic acid (PABA) and p-aminohippuric acid (PAH) via an azo bond that was cleaved by the azoreductase activities of colonic bacteria. The novel active molecule, 4-amino-THC, was readily released from the conjugates in the colon (>62% within 24h) with only very small amounts released in the upper GI tract (<12% over 12h). The polymer conjugates showed chemical stability at various pH values along the gastrointestinal tract and increased water solubility of up to 5mg/mL. 4-Amino-THC demonstrated cytotoxic ability against the human colon adenocarcinoma cell lines (HT-29) with an IC50 of 28.67 ± 1.01 μg/mL, and even greater selectivity (∼ 4 folds) to inhibit HT-29 cells than to normal human colon epithelial cell lines while curcumin was a non-selective agent against both cell lines. Our study has demonstrated that the use of THC-CMC conjugates may be a promising colon-specific drug delivery system with its sustained release in the colon to be an effective treatment for colonic cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethyl-carbodiimid, ≥97.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
N-Boc-1,6-hexandiamin, ≥98.0% (NT)