Merck
  • In vivo administration of granulocyte colony-stimulating factor restores long-term depression in hippocampal slices prepared from transgenic APP/PS1 mice.
In vivo administration of granulocyte colony-stimulating factor restores long-term depression in hippocampal slices prepared from transgenic APP/PS1 mice.
Journal of neuroscience research (2014-03-26)
Shijie Song, Xinming Wang, Vasyl Sava, Edwin J Weeber, Juan Sanchez-Ramos
ZUSAMMENFASSUNG
Granulocyte colony-stimulating factor (G-CSF) is a hematopoietic cytokine that also possesses neurotrophic and antiapoptotic properties. G-CSF has been reported to decrease amyloid burden significantly, promote hippocampal neurogenesis, and improve spatial learning in a mouse model of Alzheimer's disease. To understand better the effects of G-CSF on hippocampal-dependent learning, the present study focused on electrophysiological correlates of neuroplasticity, long-term potentiation (LTP), and long-term depression (LTD). Two cohorts of transgenic APP/PS1 mice, with or without prior bone marrow transplantation from Tg GFP mice, were treated in vivo for 2 weeks with G-CSF or vehicle. After completion of the treatments, hippocampal slices were prepared for electrophysiological studies of LTP and LTD. LTP was induced and maintained in both G-CSF-treated and vehicle-treated groups of Tg APP/PS1. In contrast, LTD could not be induced in vehicle-treated Tg APP/PS1 mice, but G-CSF treatment restored LTD. The LTP and LTD results obtained from the cohort of bone marrow-grafted Tg APP/PS1 mice did not differ from those from nongrafted Tg APP/PS1 mice. The mechanism by which G-CSF restores LTD is not known, but it is possible that its capacity to reduce amyloid plaques results in increased soluble oligomers of amyloid-β (A-β), which in turn may facilitate LTD. This mechanism would be consistent with the recent report that soluble A-β oligomers promote LTD in hippocampal slices.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
SAFC
Natriumchlorid -Lösung, 5 M
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%