Merck
  • Region- and age-dependent reductions of hippocampal long-term potentiation and NMDA to AMPA ratio in a genetic model of Alzheimer's disease.
Region- and age-dependent reductions of hippocampal long-term potentiation and NMDA to AMPA ratio in a genetic model of Alzheimer's disease.
Neurobiology of aging (2014-08-12)
Alessandro Tozzi, Alessandra Sclip, Michela Tantucci, Antonio de Iure, Veronica Ghiglieri, Cinzia Costa, Massimiliano Di Filippo, Tiziana Borsello, Paolo Calabresi
ZUSAMMENFASSUNG
To characterize the mechanisms underlying region- and age-dependent hippocampal synaptic dysfunction in Alzheimer's disease, we used transgenic CRND8 mice, expressing the Swedish-Indiana APP mutation. In 2-month-old mice, no β-amyloid plaques deposition, but the presence of soluble oligomers, were found in CA1 area but not in dentate gyrus (DG). At this age, long-term potentiation (LTP) was reduced selectively in CA1. In 6-month-old mice, the presence of soluble oligomers was accompanied by accumulation of β-amyloid plaques and decreased LTP in CA1 and DG regions. In both regions, the loss of LTP was linked to reduced N-methyl-D-aspartate (NMDA) to alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) current ratio. The acetylcholine-esterase inhibitor, neostigmine rescued LTP in CA1 area at early stage of the disease but not after plaques deposition. Conversely, the NMDA receptor antagonist memantine restored LTP selectively in DG at later stages of the disease. Both these effects were associated with a normalization of the NMDA to AMPA ratio. The association between the recovery of LTP and the normalization of the NMDA to AMPA ratio provides information on new possible therapeutic strategies in Alzheimer's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
SAFC
BIS-TRIS
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
(Aminomethyl)phosphonsäure, 99%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Supelco
(Aminomethyl)phosphonsäure, PESTANAL®, analytical standard
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis