Merck
  • Folate and TAT peptide co-modified liposomes exhibit receptor-dependent highly efficient intracellular transport of payload in vitro and in vivo.
Folate and TAT peptide co-modified liposomes exhibit receptor-dependent highly efficient intracellular transport of payload in vitro and in vivo.
Pharmaceutical research (2014-05-27)
Yaqin Zhu, Liang Cheng, Lifang Cheng, Fazhen Huang, Qing Hu, Ling Li, Chenmin Tian, Lin Wei, Dawei Chen
ZUSAMMENFASSUNG
Using different chain lengths of PEG as linkers to develop a novel folate (FA) and TAT peptide co-modified doxorubicin (DOX)-loaded liposome (FA/TAT-LP-DOX) and evaluate its potential for tumor targeted intracellular drug delivery. FA/TAT-LP-DOX was prepared by pH gradient method and post-insertion method and the optimal ligand density was screened by MTT assay. In vitro evaluation was systematically performed through cytotoxicity assay, cellular uptake studies, subcellular localization and cellular uptake mechanism in folate receptor (FR) over-expressing KB tumor cells. In vivo tumor targeted delivery of FA/TAT-LP-DOX was also studied by in vivo fluorescence imaging in a murine KB xenograft model. The particle size and zeta potential determination indicated that FA and TAT were successfully inserted into the liposome and cationic TAT peptide was completely shielded. With the optimal ligand density (5% of FA and 2.5% TAT), the FA/TAT-LP-DOX exhibited improved cytotoxity and cellular uptake efficiency compared with its single-ligand counterparts (FA-LP-DOX and PEG/TAT-LP-DOX). Competitive inhibition and uptake mechanism experiments revealed that FA and TAT peptide played a synergistic effect in facilitating intracellular transport of the liposome, and association between FA and FA receptors activated this transport process. In vivo imaging further demonstrated the superiority of FA/TAT-LP in tumor targeting and accumulation. Folate and TAT peptide co-modified liposome using different chain lengths of PEG as linkers may provide a useful strategy for specific and efficient intracellular drug delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Doxorubicin -hydrochlorid, 98.0-102.0% (HPLC)
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
Colchicin, ≥95% (HPLC), powder
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Doxorubicin -hydrochlorid, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Colchicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Doxorubicin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
Bernsteinsäure bis(N-Hydroxysuccinimid-Ester), ≥95%, powder
Sigma-Aldrich
DL-Cystein, technical grade
USP
Doxorubicin -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, puriss., ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
Colchicin, (European Pharmacopoeia (EP) Reference Standard)