Merck
  • To study physical compatibility between dibasic calcium phosphate and cohesive actives using powder rheometer and thermal methods.
To study physical compatibility between dibasic calcium phosphate and cohesive actives using powder rheometer and thermal methods.
Drug development and industrial pharmacy (2013-10-09)
Maitri R Trivedi, Rutesh H Dave
ZUSAMMENFASSUNG
An attempt is made to provide better understanding of the compatibility aspect of excipients with different properties of active pharmaceutical ingredient (API) using various rheological, thermal and morphological studies conducted on binary mixtures of Dibasic Calcium Phosphate anhydrous (DCP-A) and Dibasic Calcium Phosphate dihydrate (DCP-D) forms with cohesive API's (Acetaminophen and Aspirin). Binary mixtures of DCP's were prepared by addition of 0% w/w to 50% w/w of the API in each powder blend. Powder rheological analysis were conducted using FT4 powder rheometer, rotational shear cell and empirical approaches such as angle of repose (AOR), Hausner ratio (HR) and Carr's index (CI). Thermal analysis was conducted using differential scanning calorimetry (DSC) and thermal effusivity. Morphological studies were conducted using Scanning Electron Microscopy (SEM) to determine the fundamental differences between powder materials. Result/Conclusions: Powder rheometer showed distinctive understanding in the flowability behavior of binary mixtures with addition of increasing proportion of API's than empirical approaches. Thermal approaches revealed the potential interaction of water of crystallization DCP-D while such interaction was absent in DCP-A. Binary mixtures prepared using DCP-D were better flowable while blends containing DCP-A were better in stability (physical), compressibility and permeability. This study allows the scientist to understand the powder packing of different blends with their flowability and compressibility parameter and helps selection of appropriate form of hydrate for relevant API.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetylsalicylsäure, ≥99.0%
Supelco
Aspirin (Acetylsalicylsäure), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aspirin, meets USP testing specifications
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetylsalicylsäure, analytical standard
Acetylsalicylsäure, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylsäure für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard