Merck
  • Partial filling affinity capillary electrophoresis including adsorption energy distribution calculations--towards reliable and feasible biomolecular interaction studies.
Partial filling affinity capillary electrophoresis including adsorption energy distribution calculations--towards reliable and feasible biomolecular interaction studies.
The Analyst (2015-03-10)
Joanna Witos, Jörgen Samuelsson, Geraldine Cilpa-Karhu, Jari Metso, Matti Jauhiainen, Marja-Liisa Riekkola
ZUSAMMENFASSUNG
In this work, a method to study and analyze the interaction data in free solution by exploiting partial filling affinity capillary electrophoresis (PF-ACE) followed by adsorption energy distribution calculations (AED) prior model fit to adsorption isotherms will be demonstrated. PF-ACE-AED approach allowed the possibility to distinguish weak and strong interactions of the binding processes between the most common apolipoprotein E protein isoforms (apoE2, apoE3, apoE4) of high density lipoprotein (HDL) and apoE-containing HDL2 with major glycosaminoglycan (GAG) chain of proteoglycans (PGs), chondroitin-6-sulfate (C6S). The AED analysis clearly revealed the heterogeneity of the binding processes. The major difference was that they were heterogeneous with two different adsorption sites for apoE2 and apoE4 isoforms, whereas interestingly for apoE3 and apoE-containing HDL2, the binding was homogeneous (one site) adsorption process. Moreover, our results allowed the evaluation of differences in the binding process strengths giving the following order with C6S: apoE-containing HDL2 > apoE2 > apoE4 > apoE3. In addition, the affinity constant values determined could be compared with those obtained in our previous studies for the interactions between apoE isoforms and another important GAG chain of PGs - dermatan sulfate (DS). The success of the combination of AED calculations prior to non-linear adsorption isotherm model fit with PF-ACE when the concentration range was extended, confirmed the power of the system in the clarification of the heterogeneity of biological processes studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Supelco
Natronlauge, 49-51% in water, eluent for IC
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC