Merck
Systematic profiling of indole-3-acetic acid biosynthesis in bacteria using LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-10)
Guang-Huey Lin, Chung-Yu Chang, Huei-Ru Lin
ZUSAMMENFASSUNG
Indole-3-acetic acid (IAA) is produced from tryptophan through five synthesis pathways. A comprehensive method for the quantification of IAA and biosynthesis-related intermediates in a culture medium was developed. Sample preparation was simple with protein precipitation. The analytes were separated on a superficially porous C18 silica column and detected by electrospray ionization-tandem mass spectrometry in the positive ion multiple reaction monitoring mode. The limit of detection was 0.05 μM, and the lower limits of quantification ranged from 0.05 to 2 μM. The intra-day and inter-day precision and accuracy were less than 13.96%. Ion suppression was observed, and the deuterated internal standards were used to compensate for the matrix effect. The method was applied to analyze changes in tryptophan catabolism in a culture medium of Pseudomonas putida. The proposed method is robust and suitable for the systematic profiling of IAA biosynthesis in culture supernatant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
3-Indolessigsäure, 98%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
DL-3-(3-Indolyl)-milchsäure, 99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3-Indolessigsäure, suitable for plant cell culture, crystalline
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Indol-3-essigsäure Natriumsalz, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
3-Indolacetonitril, 98%
Supelco
3-Indolessigsäure, PESTANAL®, analytical standard
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)