Merck
Acanthamoeba T4, T5 and T11 isolated from mineral water bottles in southern Brazil.
Current microbiology (2014-08-15)
Vinicius José Maschio, Fernanda Chies, Ana Maris Carlesso, Amanda Carvalho, Sayonara Peixoto Rosa, Sueli Teresinha Van Der Sand, Marilise Brittes Rott
ZUSAMMENFASSUNG
Acanthamoeba is a protist potential pathogen, capable of causing a blinding keratitis in contact lens wearers and disseminated infection, leading to granulomatous amebic encephalitis in immunocompromised individuals. This amoeba is a ubiquitous organism that has been isolated from various domestic water systems, such as cooling towers and hospital water networks. The objective of this work was to investigate the presence of Acanthamoeba in mineral water bottles marketed in Porto Alegre, southern Brazil. Positive samples were further classified at the genotype level after sequencing the ASA.S1 region of 18S rDNA gene. Six of the eight isolates belonged to T5 genotype, one to T4 genotype, and one was T11. Several genotypes have been reported worldwide as causative of pathologies in humans, including genotypes T4, T5 and T11. Overall, the widespread distribution of potentially pathogenic Acanthamoeba strains in the studied source demands more awareness within the public and health professionals, because this pathogen is emerging as a risk for human health worldwide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O