Merck
  • Endospanin 1 silencing in the hypothalamic arcuate nucleus contributes to sustained weight loss of high fat diet obese mice.
Endospanin 1 silencing in the hypothalamic arcuate nucleus contributes to sustained weight loss of high fat diet obese mice.
Gene therapy (2014-05-03)
V Vauthier, T D Swartz, P Chen, C Roujeau, M Pagnon, J Mallet, C Sarkis, R Jockers, J Dam
ZUSAMMENFASSUNG
Leptin targets specific receptors (OB-R) expressed in the hypothalamus to regulate energy balance. Leptin decreases food intake in normal weight individuals, but this effect is blunted in obese subjects who are characterized by a state of leptin resistance. The prevention of leptin resistance is one of the major goals of obesity research. Recently, we identified endospanin 1 as a negative regulator of OB-R, which by interacting with OB-R retains the receptor inside the cell. We show here that in obese mice endospanin 1 is upregulated in the hypothalamic arcuate nucleus (ARC), the major brain structure involved in body weight regulation, suggesting that endospanin 1 is implicated in obesity development and/or the installation of leptin resistance. In contrast, silencing of endospanin 1 with lentiviral vectors in the ARC of obese mice fully restores leptin responsiveness when combined with a switch to ad libitum fed chow diet. The recovery of central leptin sensitivity is accompanied by sustained body weight loss and amelioration of blood lipid parameters and steatosis. Collectively, our results define endospanin 1 as a novel therapeutic target against obesity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.