Merck
  • Effect of exercise programs with aerobic exercise sessions of similar intensity but different frequency and duration on health-related measures in overweight women.
Effect of exercise programs with aerobic exercise sessions of similar intensity but different frequency and duration on health-related measures in overweight women.
Journal of physical activity & health (2014-06-05)
Eirini Manthou, Jason M Gill, Dalia Malkova
ZUSAMMENFASSUNG
This study investigated health-related effects of exercise programs with exercise sessions of similar intensity but different frequency and duration. Thirty-four overweight women were randomized into either long-bout (LB) or short-bout (SB) exercise groups. Participants performed an 8-week supervised program, with the LB group exercising for 75 minutes twice per week, and the SB group for 30 minutes on 5 days of the week. The LB group completed 16 sessions at a heart rate (HR) of 127 ± 1 beat·min-1 and the SB group completed 40 sessions at a HR of 126 ± 1 beat·min-1. Weekly energy expenditure of exercise was not different between groups (LB group, 5.64 ± 0.34 MJ; SB group, 5.83 ± 0.23 MJ). Training significantly (P < .05) increased measures of cardiorespiratory fitness, decreased waist circumference, insulin resistance, and diastolic blood pressure. The group × time interaction was not significant for any of these factors, indicating that responses to exercise training did not differ between the SB and LB groups. Health-related outcomes of exercise programs with similar energy expenditure are independent of frequency and duration of exercise sessions. This provides individuals with a degree of flexibility in exercise program planning.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.