Merck
A newly developed lubricant, chitosan laurate, in the manufacture of acetaminophen tablets.
International journal of pharmaceutics (2015-02-11)
Ahmad Bani-Jaber, Asuka Kobayashi, Kyohei Yamada, Dana Haj-Ali, Takeaki Uchimoto, Yasunori Iwao, Shuji Noguchi, Shigeru Itai
ZUSAMMENFASSUNG
To study the usefulness of chitosan laurate (CS-LA), a newly developed chitosan salt, as a lubricant, lubrication properties such as the pressure transmission ratio and ejection force were determined at different concentrations of CS-LA in tableting. In addition, tablet properties such as the tensile strength, disintegration time, and dissolution behavior, were also determined. When CS-LA was mixed at concentrations of 0.1%-3.0%, the pressure transmission ratio was increased in a concentration-dependent manner, and the value at a CS-LA concentration of 3% was equal to that of magnesium stearate (Mg-St), a widely used lubricant. Additionally, a reduction in the ejection force was observed at a concentration from 1%, proving that CS-LA has good lubrication performance. A prolonged disintegration time and decreased tensile strength, which are known disadvantages of Mg-St, were not observed with CS-LA. Furthermore, with CS-LA, retardation of dissolution of the drug from the tablets was not observed. Conjugation of CS with LA was found to be quite important for both lubricant and tablet properties. In conclusion, CS-LA should be useful as an alternative lubricant to Mg-St.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Magnesiumstearat, technical grade
Sigma-Aldrich
Laurinsäure, 98%
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecansäure, ≥99% (GC/titration)
Sigma-Aldrich
Magnesiumstearat, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Supelco
Dodecansäure, analytical standard
Supelco
5α-Androstan-17β-ol-3-on, VETRANAL®, analytical standard
Laurinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Millipore
Bifido Selektives Supplement B, suitable for microbiology