Merck
STAT3 regulates steady-state expression of synaptopodin in cultured mouse podocytes.
Molecular pharmacology (2014-11-27)
Mousa Abkhezr, Stuart E Dryer
ZUSAMMENFASSUNG
The transcription factor signal transducer and activator of transcription-3 (STAT3) is activated by proinflammatory cytokines and circulating factors in many cell types. Synaptopodin (Synpo) is a cytoskeleton regulatory protein expressed in podocyte foot processes that regulates the dynamics of actin filaments and the stability of small GTPases. Here we show that inhibition of STAT3 signaling using the small-molecule inhibitor benzo[b]thiophene,6-nitro-,1,1-dioxide (Stattic), or by STAT3 knockdown by small interfering RNA, caused a decrease in Synpo mRNA and protein in an immortalized mouse podocyte cell line. This loss of Synpo, which occurred in 30-80 minutes, was also seen after treatment with the translational inhibitor cycloheximide. The loss of Synpo protein after Stattic or cycloheximide treatment did not occur when podocytes were simultaneously exposed to 1-[N-[(l-3-trans-carboxyoxirane-2-carbonyl)-l-leucyl]amino]-4-guanidinobutane (E-64), an inhibitor of thiol proteases such as cathepsin L. Treatment with interleukin-6 (IL-6) increased tyrosine phosphorylation of STAT3 and evoked a parallel increase in Synpo levels in podocytes. The stimulatory effect of IL-6 on Synpo was completely inhibited by pretreatment with Stattic. By contrast, 30-60-minute exposure to angiotensin II (Ang II) inhibited STAT3 signaling and concurrently reduced Synpo protein levels. The Ang II-evoked loss of Synpo was prevented by E-64 but not by inhibition of calcineurin or blockade of transient receptor potential cation channels. Inhibition of STAT3 by Stattic caused marked changes in the distribution of podocyte actin filaments, and caused a nearly complete suppression of the migration of these cells in wound assays, consistent with the loss of Synpo. Stattic treatment also caused loss of RhoA protein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Benzoesäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Supelco
Benzoesäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
E-64, protease inhibitor
Supelco
Mettler-Toledo Kalibriersubstanz ME 18555, Benzoesäure, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Schmelzpunktstandard 121-123°C, analytical standard
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
Benzoesäure, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Millipore
Cycloheximid -Lösung, 0.1%, suitable for microbiology
USP
Benzoesäure, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzoesäure, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoesäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
Benzoesäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
Benzoesäure, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzoesäure, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoesäure, purified by sublimation, ≥99%
Sigma-Aldrich
(±)-CPP, solid
Supelco
Actidion, PESTANAL®, analytical standard
Benzoesäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
S3I-201, ≥97% (HPLC)