Merck
Antioxidant and anti-inflammatory activity of Potentilla reptans L.
Acta poloniae pharmaceutica (2015-04-09)
Marina T Tomovic, Snezana M Cupara, Marija T Popovic-Milenkovic, Biljana T Ljujic, Marina J Kostic, Slobodan M Jankovic
ZUSAMMENFASSUNG
Potentilla species have been used in traditional medicine in the treatment of different ailment, disease or malady. Potentilla reptans (P. reptans) has been scarcely studied. The aim of this study was to test antioxidant and anti-inflammatory activity of P. reptans aerial part and rhizome. DPPH assay was used to measure antioxidant activity of aqueous plant extracts. Anti-inflammatory effect was evaluated by experimental animal model of phenol-in-acetone induced mice ear edema. DPPH radical-scavenging activity of both tested extracts was concentration dependent with IC50 values 12.11 μg/mL (aerial part) and 2.57 μg/mL (rhizome). Maximum anti-inflammatory effect (61.37%) was observed after administration of 10 mg/ear of the rhizome extract and it was 89.24% of effect induced by dexamethasone as a standard. In conclusion, P. reptans rhizome aqueous extract possesses anti-inflammatory effect and higher antioxidant activity than aerial part.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Natriumcarbonat, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Natriumcarbonat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Butyliertes Hydroxytoluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
1,3-Propandiol, 98%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard