Merck
  • Clear cell myoepithelial carcinoma of salivary glands showing EWSR1 rearrangement: molecular analysis of 94 salivary gland carcinomas with prominent clear cell component.
Clear cell myoepithelial carcinoma of salivary glands showing EWSR1 rearrangement: molecular analysis of 94 salivary gland carcinomas with prominent clear cell component.
The American journal of surgical pathology (2015-01-13)
Alena Skálová, Ilan Weinreb, Martin Hyrcza, Roderick H W Simpson, Jan Laco, Abbas Agaimy, Marina Vazmitel, Hanna Majewska, Tomas Vanecek, Peter Talarčik, Spomenka Manajlovic, Simona N Losito, Petr Šteiner, Adela Klimkova, Michal Michal
ZUSAMMENFASSUNG
This study examines the presence of the EWSR1 rearrangement in a variety of clear cell salivary gland carcinomas with myoepithelial differentiation. A total of 94 salivary gland carcinomas with a prominent clear cell component included 51 cases of clear cell myoepithelial carcinomas de novo (CCMC), 21 cases of CCMCs ex pleomorphic adenoma (CCMCexPA), 11 cases of epithelial-myoepithelial carcinoma (EMC), 6 cases of EMC with solid clear cell overgrowth, and 5 cases of hyalinizing clear cell carcinoma of minor salivary glands. In addition, 10 cases of myoepithelial carcinomas devoid of clear cell change and 12 cases of benign myoepithelioma were included as well. All the tumors in this spectrum were reviewed, reclassified, and tested by fluorescence in situ hybridization (FISH) for the EWSR1 rearrangement using the Probe Vysis EWSR1 Break Apart FISH Probe Kit. The EWSR1 rearrangement was detected in 20 of 51 (39%) cases of CCMC, in 5 of 21 (24%) cases of CCMCexPA, in 1 of 11 (9%) cases of EMC, and in 4 of 5 (80%) cases of hyalinizing clear cell carcinoma. The 25 EWSR1-rearranged CCMCs and CCMCexPAs shared similar histomorphology. They were arranged in nodules composed of compact nests of large polyhedral cells with abundant clear cytoplasm. Necrosis, areas of squamous metaplasia, and hyalinization were frequent features. Immunohistochemically, the tumors expressed p63 (96%), cytokeratin CK14 (96%), and S100 protein (88%). MIB1 index varied from 10% to 100%, with most cases in the 20% to 40% range. Clinical follow-up information was available in 21 cases (84%) and ranged from 3 months to 15 years (mean 5.2 y); 4 patients were lost to follow-up. Ten patients are alive with no evidence of recurrent or metastatic disease in the follow-up period from 3 months to 15 years (mean 5 y), 3 patients are alive with recurrent and metastatic disease, and 8 died of disseminated cancer 9 months to 16 years after diagnosis (mean 6 y). Lymph node metastasis appeared in 5 patients within 5 months to 4 years after diagnosis (mean 22 mo), distant metastases were noted in 7 patients with invasion of orbit (2 cases), and in 1 case each metastasis to the neck soft tissues, liver, lungs, mediastinum, and thoracic vertebra was noted. We describe for the first time EWSR1 gene rearrangement in a subset of myoepithelial carcinomas arising in minor and major salivary glands. The EWSR1-rearranged CCMC represents a distinctive aggressive variant composed predominantly of clear cells with frequent necrosis. Most EWSR1-rearranged CCMCs of salivary glands are characterized by poor clinical outcomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98.0%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
2-Phenylindol, technical grade, 95%