Merck
  • Combining in vitro and in silico methods for better prediction of surfactant effects on the absorption of poorly water soluble drugs-a fenofibrate case example.
Combining in vitro and in silico methods for better prediction of surfactant effects on the absorption of poorly water soluble drugs-a fenofibrate case example.
International journal of pharmaceutics (2014-07-06)
Ragna Berthelsen, Erik Sjögren, Jette Jacobsen, Jakob Kristensen, René Holm, Bertil Abrahamsson, Anette Müllertz
ZUSAMMENFASSUNG
The aim of this study was to develop a sensitive and discriminative in vitro-in silico model able to simulate the in vivo performance of three fenofibrate immediate release formulations containing different surfactants. In addition, the study was designed to investigate the effect of dissolution volume when predicting the oral bioavailability of the formulations. In vitro dissolution studies were carried out using the USP apparatus 2 or a mini paddle assembly, containing 1000 mL or 100mL fasted state biorelevant medium, respectively. In silico simulations of small intestinal absorption were performed using the GI-Sim absorption model. All simulation runs were performed twice adopting either a total small intestinal volume of 533 mL or 105 mL, in order to examine the implication of free luminal water volumes for the in silico predictions. For the tested formulations, the use of a small biorelevant dissolution volume was critical for in vitro-in silico prediction of drug absorption. Good predictions, demonstrating rank order in vivo-in vitro-in silico correlations for Cmax, were obtained with in silico predictions utilizing a 105 mL estimate for the human intestinal water content combined with solubility and dissolution data performed in a mini paddle apparatus with 100mL fasted state simulated media.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Maleinsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%