Merck
Massive diversification in aging colonies of Escherichia coli.
Journal of bacteriology (2014-07-02)
Claude Saint-Ruf, Meriem Garfa-Traoré, Valérie Collin, Corinne Cordier, Christine Franceschi, Ivan Matic
ZUSAMMENFASSUNG
The evolutionary success of bacteria depends greatly on their capacity to continually generate phenotypic diversity. Structured environments are particularly favorable for diversification because of attenuated clonal interference, which renders selective sweeps nearly impossible and enhances opportunities for adaptive radiation. We examined at the microscale level the emergence and the spatial and temporal dynamics of phenotypic diversity and their underlying causes in Escherichia coli colonies. An important dynamic heterogeneity in the growth, metabolic activity, morphology, gene expression patterns, stress response induction, and death patterns among cells within colonies was observed. Genetic analysis indicated that the phenotypic variation resulted mostly from mutations and that indole production, oxidative stress, and the RpoS-regulated general stress response played an important role in the generation of diversity. We observed the emergence and persistence of phenotypic variants within single colonies that exhibited variable fitness compared to the parental strain. Some variants showed improved capacity to produce biofilms, whereas others were able to use different nutrients or to tolerate antibiotics or oxidative stress. Taken together, our data show that bacterial colonies provide an ecological opportunity for the generation and maintenance of vast phenotypic diversity, which may increase the probability of population survival in unpredictable environments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Indol, ≥99%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Dihydrorhodamin 123, ≥95%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Indol, ≥99%, FG
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis