Merck
  • STC1 interference on calcitonin family of receptors signaling during osteoblastogenesis via adenylate cyclase inhibition.
STC1 interference on calcitonin family of receptors signaling during osteoblastogenesis via adenylate cyclase inhibition.
Molecular and cellular endocrinology (2015-01-17)
Silvia R Terra, João Carlos R Cardoso, Rute C Félix, Leo Anderson M Martins, Diogo Onofre G Souza, Fatima C R Guma, Adelino Vicente M Canário, Vanessa Schein
ZUSAMMENFASSUNG
Stanniocalcin 1 (STC1) and calcitonin gene-related peptide (CGRP) are involved in bone formation/remodeling. Here we investigate the effects of STC1 on functional heterodimer complex CALCRL/RAMP1, expression and activity during osteoblastogenesis. STC1 did not modify CALCRL and ramp1 gene expression during osteoblastogenesis when compared to controls. However, plasma membrane spatial distribution of CALCRL/RAMP1 was modified in 7-day pre-osteoblasts exposed to either CGRP or STC1, and both peptides induced CALCRL and RAMP1 assembly. CGRP, but not STC1 stimulated cAMP accumulation in 7-day osteoblasts and in CALCRL/RAMP1 transfected HEK293 cells. Furthermore, STC1 inhibited forskolin stimulated cAMP accumulation of HEK293 cells, but not in CALCRL/RAMP1 transfected HEK293 cells. However, STC1 inhibited cAMP accumulation in calcitonin receptor (CTR) HEK293 transfected cells stimulated by calcitonin. In conclusion, STC1 signals through inhibitory G-protein modulates CGRP receptor spatial localization during osteoblastogenesis and may function as a regulatory factor interacting with calcitonin peptide members during bone formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Sigma-Aldrich
Carbamoylcholinchlorid, ≥98% (titration), crystalline
Sigma-Aldrich
Calcitonin Gene Related Peptide human, ≥95% (HPLC), powder
Sigma-Aldrich
Calcitonin, human, ≥97% (HPLC), powder
Supelco
Forskolin, analytical standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Carbachol, European Pharmacopoeia (EP) Reference Standard