Merck
  • Residual structure of Streptococcus mutans biofilm following complete disinfection favors secondary bacterial adhesion and biofilm re-development.
Residual structure of Streptococcus mutans biofilm following complete disinfection favors secondary bacterial adhesion and biofilm re-development.
PloS one (2015-01-31)
Tatsuya Ohsumi, Shoji Takenaka, Rika Wakamatsu, Yuuki Sakaue, Naoki Narisawa, Hidenobu Senpuku, Hayato Ohshima, Yutaka Terao, Takashi Okiji
ZUSAMMENFASSUNG
Chemical disinfection of oral biofilms often leaves biofilm structures intact. This study aimed to examine whether the residual structure promotes secondary bacterial adhesion. Streptococcus mutans biofilms generated on resin-composite disks in a rotating disc reactor were disinfected completely with 70% isopropyl alcohol, and were again cultured in the same reactor after resupplying with the same bacterial solution. Specimens were subjected to fluorescence confocal laser scanning microscopy, viable cell counts and PCR-Invader assay in order to observe and quantify secondarily adhered cells. Fluorescence microscopic analysis, particularly after longitudinal cryosectioning, demonstrated stratified patterns of viable cells on the disinfected biofilm structure. Viable cell counts of test specimens were significantly higher than those of controls, and increased according to the amount of residual structure and culture period. Linear regression analysis exhibited a high correlation between viable and total cell counts. It was concluded that disinfected biofilm structures favored secondary bacterial adhesion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles